Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL137642.1 | - | 3 | 536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000445784.1 | - | 3 | 518 | Chr. 7 - 90312496 - 90322592 [+] on Build GRCh38 |