Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103446.1 | 3 | 1656 | |||
NR_103447.1 | 4 | 1162 | ||||
GenBank mRNA | AK095117.1 | - | 3 | 1551 | ||
CX787946.1 | - | 2 | 296 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000606714.5 | - | 3 | 1551 | Chr. 6 - 52577307 - 52583985 [+] on Build GRCh38 |