Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103446.1 | 1 | 799 | |||
GenBank mRNA | AK095117.1 | - | 1 | 694 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000606714.5 | - | 1 | 694 | Chr. 6 - 52577307 - 52583985 [+] on Build GRCh38 |