Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101339.2 | NP_001094809.2 | 3 | 566 | ||
XM_017018966.1 | XP_016874455.1 | 4 | 577 | |||
GenBank mRNA | BC035880.1 | - | 3 | 577 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000623395.3 | - | 566 | Chr. - - [-] on Build |