Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120519.1 | 3 | 300 | |||
NR_120520.1 | 3 | 277 | ||||
GenBank mRNA | AA025193.1 | - | 3 | 252 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000448513.1 | - | 3 | 270 | Chr. 7 - 101014325 - 101017601 [-] on Build GRCh38 |