Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110291.1 | 3 | 220 | |||
GenBank mRNA | AK093026.1 | - | 2 | 876 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000452057.1 | - | 2 | 877 | Chr. 2 - 210030733 - 210064356 [+] on Build GRCh38 |