Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027098.1 | 5 | 717 | |||
NR_027100.1 | 4 | 454 | ||||
GenBank mRNA | AK095182.1 | - | 5 | 717 | ||
BC121144.1 | - | 4 | 454 | |||
BC121145.1 | - | 4 | 457 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435075.1 | - | 4 | 454 | Chr. 2 - 32882801 - 32926522 [+] on Build GRCh38 |