Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027098.1 | 5 | 1407 | |||
NR_027100.1 | 4 | 1144 | ||||
GenBank mRNA | AK095182.1 | - | 5 | 1407 | ||
BC121144.1 | - | 4 | 1144 | |||
BC121145.1 | - | 4 | 1147 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435075.1 | - | 4 | 1144 | Chr. 2 - 32882801 - 32926522 [+] on Build GRCh38 |