Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRERNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051976.1 | 2 | 525 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000431460.1 | - | 2 | 400 | Chr. 20 - 50040716 - 50041504 [-] on Build GRCh38 |