Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110259.1 | 5 | 1005 | |||
GenBank mRNA | BG219532.1 | - | 2 | 307 |