Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110115.1 | 2 | 503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419422.1 | - | 2 | 497 | Chr. 7 - 101308346 - 101310985 [+] on Build GRCh38 |