Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110699.1 | 4 | 1290 | |||
GenBank mRNA | AK092140.1 | - | 4 | 1291 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000415437.1 | - | 4 | 1290 | Chr. 1 - 163161675 - 163213023 [+] on Build GRCh38 |