Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110224.1 | 5 | 1007 | |||
GenBank mRNA | AK097952.1 | - | 5 | 1002 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000445865.1 | - | 5 | 1002 | Chr. 2 - 64846130 - 64863626 [-] on Build GRCh38 |