Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100130548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135126.1 | 1, 2 | 196 | |||
GenBank mRNA | AK057520.1 | - | 1, 2 | 196 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000412181.1 | - | 1, 2 | 196 | Chr. 9 - 134054290 - 134058805 [+] on Build GRCh38 |