Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: LOC100507477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121622.1 | 4 | 915 | |||
NR_121623.1 | 2 | 396 | ||||
GenBank mRNA | BC038188.1 | - | 4 | 904 | ||
BU160297.1 | - | 2 | 396 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446458.1 | - | 2 | 362 | Chr. 6 - 140086682 - 140093665 [+] on Build GRCh38 |