Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELFN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120508.1 | 1 | 91 | |||
NR_120509.1 | 1 | 91 | ||||
NR_120510.1 | 1 | 91 | ||||
GenBank mRNA | BF665480.1 | - | 73 | |||
HO663747.1 | - | 1 | 74 | |||
HY180852.1 | - | 1 | 92 | |||
JX046910.1 | - | 1 | 74 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000453348.1 | - | 1 | 53 | Chr. 7 - 1738630 - 1742272 [-] on Build GRCh38 |