Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037881.1 | 3 | 2194 | |||
GenBank mRNA | BC043283.1 | - | 3 | 1841 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000454455.5 | - | 3 | 2232 | Chr. 2 - 3957655 - 3974070 [-] on Build GRCh38 |