Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101337.1 | NP_001094807.1 | 1 | 154 | ||
GenBank mRNA | AF086445.1 | - | 1 | 154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000623038.1 | - | 154 | Chr. - - [-] on Build |