Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf67-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110820.1 | 3 | 1905 | |||
GenBank mRNA | AK090895.1 | - | 3 | 1905 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000482372.1 | - | 3 | 1905 | Chr. 3 - 58824471 - 59019093 [+] on Build GRCh38 |