- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Gene Symbol: LINC01024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102739.1 | 3 | 2424 | |||
NR_102740.1 | 4 | 1787 | ||||
NR_102741.1 | 4 | 1506 | ||||
GenBank mRNA | BC042448.1 | - | 3 | 2055 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499203.6 | - | 3 | 2055 | Chr. 5 - 140102922 - 140107643 [-] on Build GRCh38 |