Gene Symbol: LINC01024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102739.1 | 1 | 1361 | |||
GenBank mRNA | BC042448.1 | - | 1 | 992 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499203.6 | - | 1 | 992 | Chr. 5 - 140102922 - 140107643 [-] on Build GRCh38 |