Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100287015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040040.1 | 2 | 2325 | |||
GenBank mRNA | AK025595.1 | - | 2 | 2323 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000500118.2 | - | 2 | 2325 | Chr. 8 - 6403551 - 6406548 [-] on Build GRCh38 |