Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDNF-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103442.1 | 1 | 1486 | |||
GenBank mRNA | JF824131.1 | - | 1 | 1486 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000514532.2 | - | 1 | 1486 | Chr. 5 - 37869907 - 37874894 [+] on Build GRCh38 |