Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125919.1 | 4 | 1411 | |||
GenBank mRNA | BC029598.1 | - | 4 | 1413 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000509964.1 | - | 4 | 1413 | Chr. 4 - 17171757 - 17186059 [-] on Build GRCh38 |