Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI459905.1 | - | 2 | 147 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000522778.1 | - | 2 | 142 | Chr. 8 - 111183292 - 111236203 [-] on Build GRCh38 |