Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516833.2 | XP_011515135.1 | 1 | 809 | ||
XM_011516834.2 | XP_011515136.1 | 1 | 809 | |||
XM_011516836.2 | XP_011515138.1 | 1 | 809 | |||
XM_011516837.2 | XP_011515139.1 | 1 | 809 | |||
XM_011516838.2 | XP_011515140.1 | 1 | 809 | |||
XM_011516839.2 | XP_011515141.1 | 1 | 809 | |||
XM_011516840.2 | XP_011515142.1 | 1 | 809 | |||
XM_011516841.2 | XP_011515143.1 | 1 | 809 | |||
XM_011516842.2 | XP_011515144.1 | 1 | 809 | |||
XM_017013029.1 | XP_016868518.1 | 1 | 809 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000523905.5 | - | 1 | 296 | Chr. 8 - 94950037 - 94951396 [+] on Build GRCh38 |