Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130737.1 | 7 | 1543 | |||
GenBank mRNA | AK126009.1 | - | 6 | 3063 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000522615.1 | - | 6 | 3072 | Chr. 5 - 168706567 - 168720884 [+] on Build GRCh38 |