Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 1767 | |||
NR_137175.1 | 2 | 2038 | ||||
NR_137176.1 | 2 | 1749 | ||||
NR_137177.1 | 2 | 1728 | ||||
NR_137179.1 | 5 | 1721 | ||||
NR_137180.1 | 5 | 1779 | ||||
NR_137181.1 | 3 | 1664 | ||||
NR_137182.1 | 4 | 1746 | ||||
NR_137183.1 | 3 | 1655 | ||||
NR_137184.1 | 2 | 1573 | ||||
NR_137185.1 | 3 | 1634 | ||||
NR_137186.1 | 3 | 1543 | ||||
NR_137187.1 | 3 | 1522 | ||||
NR_137188.1 | 3 | 1612 | ||||
NR_137189.1 | 2 | 2059 | ||||
NR_137190.1 | 3 | 2092 | ||||
NR_137191.1 | 3 | 2071 | ||||
NR_137192.1 | 4 | 1931 | ||||
NR_137193.1 | 5 | 1758 | ||||
NR_137194.1 | 3 | 1633 | ||||
NR_137195.1 | 2 | 1801 | ||||
NR_137196.1 | 2 | 1822 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 2291 | ||
AK021552.1 | - | 1 | 933 | |||
AK123947.1 | - | 2 | 1975 | |||
AL832469.1 | - | 3 | 1327 | |||
BC040599.1 | - | 2 | 1351 | |||
BX538058.1 | - | 2 | 1329 | |||
BX647608.1 | - | 2 | 1329 |