Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC387810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104155.1 | 4 | 1001 | |||
GenBank mRNA | BC070394.1 | - | 4 | 1001 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000525984.2 | - | 4 | 1001 | Chr. 11 - 112534222 - 112555802 [-] on Build GRCh38 |