Gene Symbol: LOC100996671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110036.1 | 2 | 306 | |||
GenBank mRNA | BX111310.1 | - | 2 | 256 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000537478.1 | - | 2 | 256 | Chr. 12 - 126663679 - 126690268 [-] on Build GRCh38 |