Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105370622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135190.1 | 2 | 850 | |||
GenBank mRNA | BC039357.1 | - | 2 | 850 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553712.1 | - | 2 | 850 | Chr. 14 - 90697333 - 90699374 [+] on Build GRCh38 |