Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135286.1 | 2 | 3021 | |||
GenBank mRNA | AK125955.1 | - | 2 | 3022 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000554257.1 | - | 2 | 3021 | Chr. 14 - 37896060 - 37902124 [-] on Build GRCh38 |