Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134922.1 | 2 | 927 | |||
NR_134923.1 | 2 | 904 | ||||
NR_134924.1 | 2 | 837 | ||||
GenBank mRNA | AK057319.1 | - | 2 | 927 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000566649.1 | - | 2 | 1134 | Chr. 16 - 54366007 - 54370665 [-] on Build GRCh38 |