Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136400.1 | 1 | 373 | |||
GenBank mRNA | BC033940.1 | - | 1 | 376 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000574471.1 | - | 1 | 375 | Chr. 17 - 82713908 - 82716255 [-] on Build GRCh38 |