Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100505797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134647.1 | 3 | 270 | |||
NR_134648.1 | 2 | 96 | ||||
GenBank mRNA | BG182684.1 | - | 1 | 24 | ||
BX114234.1 | - | 3 | 270 |