Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D630029K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027846.1 | 1 | 110 | |||
GenBank mRNA | AK085456.1 | - | 1 | 19 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA481170.1 | - | 55 | Chr. - - [-] on Build |