Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D630029K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027846.1 | 3 | 595 | |||
NR_027847.1 | 3 | 705 | ||||
GenBank mRNA | AK085456.1 | - | 3 | 504 |