Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm12522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040560.1 | 3 | 2041 | |||
GenBank mRNA | AK090264.1 | - | 3 | 2042 | ||
AK144267.1 | - | 3 | 2042 |