Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130006D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045832.1 | 5 | 890 | |||
NR_045833.1 | 5 | 710 | ||||
NR_045834.1 | 4 | 623 | ||||
GenBank mRNA | AK087393.1 | - | 5 | 866 | ||
BB537478.2 | - | 3 | 618 | |||
BC087866.1 | - | 5 | 707 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU562406.1 | - | 5 | 710 | Chr. 5 - 111734313 - 111761728 [-] on Build GRCm38 |