Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700003D09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045477.1 | 3 | 346 | |||
GenBank mRNA | AK005627.1 | - | 3 | 346 |