Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210408F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040257.1 | 8 | 968 | |||
NR_040258.1 | 7 | 714 | ||||
NR_040259.1 | 6 | 689 | ||||
NR_040260.1 | 6 | 606 | ||||
NR_040261.1 | 5 | 588 | ||||
NR_040262.1 | 6 | 553 | ||||
GenBank mRNA | AK008857.1 | - | 6 | 519 | ||
BC021953.1 | - | 8 | 875 | |||
BC099512.1 | - | 7 | 634 |