Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700007L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045709.1 | 2 | 665 | |||
GenBank mRNA | AK160141.1 | - | 2 | 657 | ||
AV039549.3 | - | 916 | ||||
BC052793.1 | - | 2 | 596 |