Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700003C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045478.1 | 3 | 323 | |||
GenBank mRNA | AK005625.1 | - | 3 | 324 |
Gene Symbol: 4930437M23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015330.1 | - | 4 | 377 |
Gene Symbol: 4930558F17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016172.1 | - | 401 |