Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2010308F09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045429.1 | 3 | 585 | |||
NR_045430.1 | 3 | 439 | ||||
GenBank mRNA | BB648418.1 | - | 3 | 387 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB363214.2 | - | 3 | 397 | Chr. X - 13234641 - 13236230 [-] on Build GRCm38 |