Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135292.1 | 5 | 1962 | |||
GenBank mRNA | BC082979.1 | - | 5 | 1769 |
Gene Symbol: NUTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540122.2 | XP_011538424.1 | 3 | 2000 | ||
XM_017016607.1 | XP_016872096.1 | 3 | 2001 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000638195.1 | - | 1776 | Chr. - - [-] on Build |