Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSER1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038337.2 | 3 | 634 | |||
NR_038338.2 | 3 | 503 | ||||
NR_038339.2 | 3 | 346 | ||||
GenBank mRNA | BC019858.1 | - | 2 | 387 | ||
BC032458.1 | - | 3 | 294 | |||
BC048277.1 | - | 3 | 346 | |||
BC067866.1 | - | 3 | 303 | |||
BC094781.1 | - | 3 | 303 | |||
BI917173.1 | - | 3 | 504 | |||
BX459928.2 | - | 3 | 632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY262147.1 | - | 1 | 260 | Chr. 20 - 44224696 - 44225137 [+] on Build GRCh38 |