Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 2524 | |||
NR_137175.1 | 2 | 2795 | ||||
NR_137176.1 | 2 | 2506 | ||||
NR_137177.1 | 2 | 2485 | ||||
NR_137179.1 | 5 | 2478 | ||||
NR_137180.1 | 5 | 2536 | ||||
NR_137181.1 | 3 | 2421 | ||||
NR_137182.1 | 4 | 2503 | ||||
NR_137183.1 | 3 | 2412 | ||||
NR_137184.1 | 2 | 2330 | ||||
NR_137185.1 | 3 | 2391 | ||||
NR_137186.1 | 3 | 2300 | ||||
NR_137187.1 | 3 | 2279 | ||||
NR_137188.1 | 3 | 2369 | ||||
NR_137189.1 | 2 | 2816 | ||||
NR_137190.1 | 3 | 2849 | ||||
NR_137191.1 | 3 | 2828 | ||||
NR_137192.1 | 4 | 2688 | ||||
NR_137193.1 | 5 | 2515 | ||||
NR_137194.1 | 3 | 2390 | ||||
NR_137195.1 | 2 | 2558 | ||||
NR_137196.1 | 2 | 2579 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 3047 | ||
AK021552.1 | - | 1 | 1692 | |||
AK091713.1 | - | 4 | 1328 | |||
AK123947.1 | - | 2 | 2731 | |||
AL832469.1 | - | 3 | 2083 | |||
BC040599.1 | - | 2 | 2109 | |||
BX538058.1 | - | 2 | 2086 | |||
BX647608.1 | - | 2 | 2087 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN345412.1 | - | 1 | 584 | Chr. 11 - 122089688 - 122090290 [-] on Build GRCh38 |