Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038464.1 | 2 | 1023 | |||
GenBank mRNA | BC009626.1 | - | 2 | 648 | ||
BC048265.1 | - | 2 | 1029 |