Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ASB16-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049729.1 | 1 | 115 | |||
NR_049730.1 | 1 | 115 | ||||
GenBank mRNA | AK055685.1 | - | 1 | 69 | ||
CB151608.1 | - | 1 | 112 | |||
CN367999.1 | - | 1 | 59 | |||
DA567080.1 | - | 1 | 115 |