Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTTY12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001551.1 | 2 | 746 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF332241.3 | - | 2 | 746 | Chr. Y - 7804924 - 7810683 [+] on Build GRCh38 |